Product Details

SNP ID
rs2107675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138338938 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCCTACCTTGTCTTGGTACAGCA[C/T]AACCTCAGCATCCCTGAAACAGGAG
Phenotype
MIM: 157680 MIM: 609372
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC25C PubMed Links

Gene Details

Gene
CDC25C
Gene Name
cell division cycle 25C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287582.1 Intron NP_001274511.1
NM_001287583.1 Intron NP_001274512.1
NM_001318098.1 Intron NP_001305027.1
NM_001790.4 Intron NP_001781.2
NM_022809.3 Intron NP_073720.1
XM_005272145.3 Intron XP_005272202.1
XM_006714739.3 Intron XP_006714802.1
XM_011543759.2 Intron XP_011542061.1
XM_011543760.2 Intron XP_011542062.1
XM_011543761.2 Intron XP_011542063.1
XM_011543762.1 Intron XP_011542064.1
XM_011543763.1 Intron XP_011542065.1
XM_011543764.1 Intron XP_011542066.1
XM_017010101.1 Intron XP_016865590.1
Gene
FAM53C
Gene Name
family with sequence similarity 53 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135647.1 Intron NP_001129119.1
NM_016605.2 Intron NP_057689.1

View Full Product Details