Product Details

SNP ID
rs2107713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107591924 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACTACACTCAGACAAATTCAGAA[C/T]GTGAGAACTTTCTTTGTGCTTATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCAP29 PubMed Links

Gene Details

Gene
BCAP29
Gene Name
B-cell receptor-associated protein 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008405.2 Intron NP_001008405.1
NM_018844.3 Intron NP_061332.2
XM_006716051.2 Intron XP_006716114.1
XM_011516406.2 Intron XP_011514708.1
XM_011516407.1 Intron XP_011514709.1
XM_011516408.2 Intron XP_011514710.1
XM_011516409.2 Intron XP_011514711.1
XM_017012436.1 Intron XP_016867925.1
XM_017012437.1 Intron XP_016867926.1
XM_017012438.1 Intron XP_016867927.1

View Full Product Details