Product Details

SNP ID
rs2091952
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119653327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACACTCATTTCAAACCACCTTT[C/T]TTAGGAGTCTGCGTGGATGTACATG
Phenotype
MIM: 603883
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
BAG3 PubMed Links

Gene Details

Gene
BAG3
Gene Name
BCL2 associated athanogene 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004281.3 Intron NP_004272.2

View Full Product Details