Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001160113.1 | 1312 | Intron | NP_001153585.1 | ||
NM_001160114.1 | 1312 | Intron | NP_001153586.1 | ||
NM_001160115.1 | 1312 | Intron | NP_001153587.1 | ||
NM_001160116.1 | 1312 | Intron | NP_001153588.1 | ||
NM_144597.2 | 1312 | Intron | NP_653198.2 | ||
XM_006720385.2 | 1312 | UTR 3 | XP_006720448.1 | ||
XM_011521212.2 | 1312 | UTR 3 | XP_011519514.1 | ||
XM_011521213.2 | 1312 | Intron | XP_011519515.1 | ||
XM_011521214.2 | 1312 | Intron | XP_011519516.1 |