Product Details
- SNP ID
-
rs2851011
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:99414826 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CTGTAAAATAAGGAAATTTTGTGGC[T/C]TACTTGAAATAAATAGCAGCTTTAT
- Phenotype
-
MIM: 600086
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADH7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs57742750] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADH7
- Gene Name
- alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide
View Full Product Details