Product Details

SNP ID
rs2136422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63477791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATACTAAAAACCAACATGCCAGG[C/T]TATTCCAGAGACTTGTAACCAGATC
Phenotype
MIM: 154790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINB5 PubMed Links

Gene Details

Gene
SERPINB5
Gene Name
serpin family B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002639.4 Intron NP_002630.2

View Full Product Details