Product Details

SNP ID
rs2829729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25397616 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGGAGAATGGAAACACTTCTACC[C/T]GGCGCATGTAGGGTACAGGTGGGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00158 PubMed Links

Gene Details

Gene
LINC00158
Gene Name
long intergenic non-protein coding RNA 158
There are no transcripts associated with this gene.

View Full Product Details