Product Details

SNP ID
rs2829888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25742555 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCATAAGAAAGAGGGAATGCATG[C/T]TTCTGTGGAATTTCATGTATAGATA
Phenotype
MIM: 603152 MIM: 600609
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5J PubMed Links

Gene Details

Gene
ATP5J
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6
There are no transcripts associated with this gene.

Gene
GABPA
Gene Name
GA binding protein transcription factor alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197297.1 Intron NP_001184226.1
NM_002040.3 Intron NP_002031.2
XM_005260938.4 Intron XP_005260995.1
XM_011529520.2 Intron XP_011527822.1
XM_011529521.2 Intron XP_011527823.1
XM_017028313.1 Intron XP_016883802.1

View Full Product Details