Product Details

SNP ID
rs2852507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:101396377 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGTACATTTAAAAAATGTCTTTA[A/G]CCACTGAATATTGACAGCATGTTAT
Phenotype
MIM: 138244
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIK2 PubMed Links

Gene Details

Gene
GRIK2
Gene Name
glutamate ionotropic receptor kainate type subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166247.1 Intron NP_001159719.1
NM_021956.4 Intron NP_068775.1
NM_175768.3 Intron NP_786944.1
XM_005266945.2 Intron XP_005267002.1
XM_005266946.3 Intron XP_005267003.1
XM_011535777.2 Intron XP_011534079.1
XM_011535778.2 Intron XP_011534080.1
XM_017010781.1 Intron XP_016866270.1
XM_017010782.1 Intron XP_016866271.1

View Full Product Details