Product Details

SNP ID
rs2852649
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:158749233 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTAAAGTTCCAATACAATTGATT[A/T]AACAATGCACCTTTTTCCACAGACC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6K6 PubMed Links

Gene Details

Gene
OR6K6
Gene Name
olfactory receptor family 6 subfamily K member 6
There are no transcripts associated with this gene.

Gene
OR6N1
Gene Name
olfactory receptor family 6 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005185.1 Intron NP_001005185.1
XM_017000325.1 Intron XP_016855814.1
XM_017000326.1 Intron XP_016855815.1
XM_017000327.1 Intron XP_016855816.1

View Full Product Details