Product Details

SNP ID
rs2146260
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39315587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTCGGCTGAGAGCTTCCTGAGG[A/G]CAGAGATCCTCTTTTCACTAAGATT
Phenotype
MIM: 607369 MIM: 607370
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KCNK16 PubMed Links
Additional Information
For this assay, SNP(s) [rs116090174] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KCNK16
Gene Name
potassium two pore domain channel subfamily K member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135105.1 Intron NP_001128577.1
NM_001135106.1 Intron NP_001128578.1
NM_001135107.1 Intron NP_001128579.1
NM_032115.3 Intron NP_115491.1
XM_011514935.2 Intron XP_011513237.1
XM_017011346.1 Intron XP_016866835.1
Gene
KCNK17
Gene Name
potassium two pore domain channel subfamily K member 17
There are no transcripts associated with this gene.

View Full Product Details