Product Details

SNP ID
rs2858196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103779577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTCTACCTCACTTCTCCCTTTG[C/G]CCGTGCAGAGCCAGAGGAGTTCTTG
Phenotype
MIM: 300401
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLP1 PubMed Links

Gene Details

Gene
PLP1
Gene Name
proteolipid protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000533.4 Intron NP_000524.3
NM_001128834.2 Intron NP_001122306.1
NM_001305004.1 Intron NP_001291933.1
NM_199478.2 Intron NP_955772.1

View Full Product Details