Product Details

SNP ID
rs2837371
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:40011216 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTACATTCTGAAACCATGGATGCA[A/C]ACCTCACATTCCTGGAGTCATCTAA
Phenotype
MIM: 602523
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DSCAM PubMed Links

Gene Details

Gene
DSCAM
Gene Name
DS cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271534.1 7364 Intron NP_001258463.1
NM_001389.3 7364 Intron NP_001380.2
XM_017028281.1 7364 UTR 3 XP_016883770.1

View Full Product Details