Product Details

SNP ID
rs2837374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:40013833 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAAGATGGACAGCTGGCTCTCGTC[C/T]TGGCAATTCTATGAAACCTTGTGAC
Phenotype
MIM: 602523
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSCAM PubMed Links

Gene Details

Gene
DSCAM
Gene Name
DS cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271534.1 Intron NP_001258463.1
NM_001389.3 Intron NP_001380.2
XM_017028281.1 Intron XP_016883770.1

View Full Product Details