Product Details
- SNP ID
-
rs2147441
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:11831048 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCACCAGGAGGTCTTGTGTGAGCTT[C/G]TTCCACCCTGGTGTGGATTTAAATC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PROSER2
PubMed Links
Gene Details
- Gene
- PROSER2
- Gene Name
- proline and serine rich 2
View Full Product Details