Product Details

SNP ID
rs2859068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47349455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTGCGAGTCCAGGGGCGAGGG[A/C]AGTCTCGAGATGAGGAGAAATGTAG
Phenotype
MIM: 609949 MIM: 615475
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C5AR2 PubMed Links

Gene Details

Gene
C5AR2
Gene Name
complement component 5a receptor 2
There are no transcripts associated with this gene.

Gene
DHX34
Gene Name
DEAH-box helicase 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014681.5 77 Intron NP_055496.2
XM_005259443.3 77 UTR 5 XP_005259500.1
XM_011527550.2 77 Intron XP_011525852.1
XM_011527551.2 77 Intron XP_011525853.1
XM_017027524.1 77 Intron XP_016883013.1

View Full Product Details