Product Details

SNP ID
rs2859167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100820758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTCTAGAGGAGAAGGCTCACGG[C/T]AACAGCTCCTCACTGTCCCGTACTA
Phenotype
MIM: 300907
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSTF2 PubMed Links

Gene Details

Gene
CSTF2
Gene Name
cleavage stimulation factor subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306206.1 Intron NP_001293135.1
NM_001306209.1 Intron NP_001293138.1
NM_001325.2 Intron NP_001316.1

View Full Product Details