Product Details

SNP ID
rs2838357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43729637 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGGAATGCACATGAGGGGAGCT[A/G]GGAGCAGAACAGTAGCACGGGCCGA
Phenotype
MIM: 179020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105372824 PubMed Links

Gene Details

Gene
LOC105372824
Gene Name
uncharacterized LOC105372824
There are no transcripts associated with this gene.

Gene
PDXK
Gene Name
pyridoxal (pyridoxine, vitamin B6) kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003681.4 Intron NP_003672.1
XM_005261195.2 Intron XP_005261252.2
XM_005261196.3 Intron XP_005261253.1
XM_005261198.2 Intron XP_005261255.1
XM_005261199.2 Intron XP_005261256.1
XM_011529758.2 Intron XP_011528060.1
XM_011529760.2 Intron XP_011528062.1
XM_011529761.1 Intron XP_011528063.1
XM_011529762.2 Intron XP_011528064.1
XM_017028483.1 Intron XP_016883972.1
XM_017028484.1 Intron XP_016883973.1

View Full Product Details