Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_003681.4 | 238 | Intron | NP_003672.1 | ||
XM_005261195.2 | 238 | Intron | XP_005261252.2 | ||
XM_005261196.3 | 238 | Intron | XP_005261253.1 | ||
XM_005261198.2 | 238 | Intron | XP_005261255.1 | ||
XM_005261199.2 | 238 | Intron | XP_005261256.1 | ||
XM_011529758.2 | 238 | Intron | XP_011528060.1 | ||
XM_011529760.2 | 238 | Intron | XP_011528062.1 | ||
XM_011529761.1 | 238 | Intron | XP_011528063.1 | ||
XM_011529762.2 | 238 | Intron | XP_011528064.1 | ||
XM_017028483.1 | 238 | Intron | XP_016883972.1 | ||
XM_017028484.1 | 238 | UTR 5 | XP_016883973.1 |