Product Details

SNP ID
rs2165570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:88007778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATCTGACACGTAGTTGCTTATTA[C/T]TCTCTCTGTGTGTATGTGTGTGTGT
Phenotype
MIM: 604620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR65 PubMed Links

Gene Details

Gene
GPR65
Gene Name
G protein-coupled receptor 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003608.3 Intron NP_003599.2

View Full Product Details