Product Details

SNP ID
rs2170656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:56815469 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTGACTTATTTTTCTCTCAAACA[A/G]TATTCCCTAATGTGGCTCCTATGAG
Phenotype
MIM: 600037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTX2 PubMed Links

Gene Details

Gene
OTX2
Gene Name
orthodenticle homeobox 2
There are no transcripts associated with this gene.

Gene
OTX2-AS1
Gene Name
OTX2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details