Product Details

SNP ID
rs2904880
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:28933075 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GATCTGGGAGGGAGAGCCTCCGTGT[C/G]TCCCACCGAGGGACAGCCTGAACCA
Phenotype
MIM: 107265 MIM: 611869
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD19 PubMed Links

Gene Details

Gene
CD19
Gene Name
CD19 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178098.1 383 Missense Mutation CTC,GTC L174V NP_001171569.1
NM_001770.5 383 Missense Mutation CTC,GTC L174V NP_001761.3
XM_006721103.3 383 Missense Mutation CTC,GTC L85V XP_006721166.1
XM_011545981.1 383 Missense Mutation CTC,GTC L174V XP_011544283.1
XM_017023893.1 383 Missense Mutation CTC,GTC L85V XP_016879382.1
Gene
RABEP2
Gene Name
rabaptin, RAB GTPase binding effector protein 2
There are no transcripts associated with this gene.

View Full Product Details