Product Details
- SNP ID
-
rs2074469
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:29440193 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTGCCCTCCAGTCCCCTATGTAC[T/C]TCTTCCTGCGCACCCTCTCGGCCTT
- Phenotype
-
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR10C1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11755177] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR10C1
- Gene Name
- olfactory receptor family 10 subfamily C member 1 (gene/pseudogene)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013941.3 |
255 |
Missense Mutation |
CTC,TTC |
L60F |
NP_039229.3 |
- Gene
- OR5V1
- Gene Name
- olfactory receptor family 5 subfamily V member 1
View Full Product Details