Product Details
- SNP ID
-
rs2074503
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:30562719 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GATCCAGCCTGTAGAGACTCGCCTT[C/T]GGGACCCATCTTTGCTTCCTTTCAG
- Phenotype
-
MIM: 603429
MIM: 143024
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78954739] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCF1
- Gene Name
- ATP binding cassette subfamily F member 1
There are no transcripts associated with this gene.
- Gene
- GNL1
- Gene Name
- G protein nucleolar 1 (putative)
There are no transcripts associated with this gene.
- Gene
- PRR3
- Gene Name
- proline rich 3
View Full Product Details