Product Details

SNP ID
rs2928834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:108609857 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCAAGCAATTGCTTAAGCATGT[A/G]TGTCCTTGTACTCAGCATAATTTAA
Phenotype
MIM: 613935
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM74 PubMed Links

Gene Details

Gene
TMEM74
Gene Name
transmembrane protein 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153015.2 Intron NP_694560.1

View Full Product Details