Product Details
- SNP ID
-
rs2911909
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:37828666 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCTTTGTTTCTCTCAGGCTACTT[A/G]GTCTTTCATAGATATCAAAGACCAA
- Phenotype
-
MIM: 172000
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PGM2
PubMed Links
Gene Details
- Gene
- PGM2
- Gene Name
- phosphoglucomutase 2
View Full Product Details