Product Details

SNP ID
rs2929182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6261327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGATAGTGGGAACACAAGTGGA[A/G]CACCTCTAGCCCAGCCCAGCCTCCT
Phenotype
MIM: 118445
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCKBR PubMed Links

Gene Details

Gene
CCKBR
Gene Name
cholecystokinin B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318029.1 Intron NP_001304958.1
NM_176875.3 Intron NP_795344.1
XM_005253210.1 Intron XP_005253267.1
XM_017018516.1 Intron XP_016874005.1

View Full Product Details