Product Details

SNP ID
rs2075592
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74410681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCACCCCCAGCCAGAGGGAGGC[A/G]GCCAGGGCACAGGCATGACCCAGCA
Phenotype
MIM: 607961
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MIR6881 PubMed Links

Gene Details

Gene
MIR6881
Gene Name
microRNA 6881
There are no transcripts associated with this gene.

Gene
SEMA7A
Gene Name
semaphorin 7A (John Milton Hagen blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146029.2 1881 Silent Mutation GCC,GCT A634A NP_001139501.1
NM_001146030.2 1881 Silent Mutation GCC,GCT A483A NP_001139502.1
NM_003612.4 1881 Silent Mutation GCC,GCT A648A NP_003603.1

View Full Product Details