Product Details

SNP ID
rs2955602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:21934974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGACATCTGTGTGATTAATGAA[A/G]AAAAAACTTCATATGTTTTCATTTG
Phenotype
MIM: 606140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XPO7 PubMed Links

Gene Details

Gene
XPO7
Gene Name
exportin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015024.4 Intron NP_055839.3
XM_017013244.1 Intron XP_016868733.1
XM_017013245.1 Intron XP_016868734.1
XM_017013246.1 Intron XP_016868735.1
XM_017013247.1 Intron XP_016868736.1

View Full Product Details