Product Details

SNP ID
rs2956110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34916568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACCTAAAGGCACCGCTAGCGT[A/C]TGGGGGCGTGGCCAACCATGCGGGA
Phenotype
MIM: 612491 MIM: 608769
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APIP PubMed Links

Gene Details

Gene
APIP
Gene Name
APAF1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015957.3 439 Intron NP_057041.2
XM_011520154.2 439 Intron XP_011518456.1
XM_017017875.1 439 Intron XP_016873364.1
Gene
PDHX
Gene Name
pyruvate dehydrogenase complex component X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135024.1 439 Intron NP_001128496.1
NM_001166158.1 439 UTR 5 NP_001159630.1
NM_003477.2 439 UTR 5 NP_003468.2
XM_011520390.1 439 Intron XP_011518692.1

View Full Product Details