Product Details

SNP ID
rs2241921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60396829 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTCAGGGAGGGAGTTAATTCTA[C/T]TTTTCCCCCTTCACCTTACATGAGA
Phenotype
MIM: 606502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MS4A14 PubMed Links

Gene Details

Gene
MS4A14
Gene Name
membrane spanning 4-domains A14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079692.2 Intron NP_001073160.1
NM_001261827.1 Intron NP_001248756.1
NM_001261828.1 Intron NP_001248757.1
NM_032597.4 Intron NP_115986.3
Gene
MS4A7
Gene Name
membrane spanning 4-domains A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021201.4 Intron NP_067024.1
NM_206938.1 Intron NP_996821.1
NM_206939.1 Intron NP_996822.1
NM_206940.1 Intron NP_996823.1

View Full Product Details