Product Details

SNP ID
rs2238833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46158704 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGGTAATAGGGATATTTAAGTGG[A/G]ATGAATTTTTTGAAGCATTTTCAGG
Phenotype
MIM: 170998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPARA PubMed Links

Gene Details

Gene
PPARA
Gene Name
peroxisome proliferator activated receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001928.2 Intron NP_001001928.1
NM_005036.4 Intron NP_005027.2
XM_005261655.3 Intron XP_005261712.1
XM_005261656.3 Intron XP_005261713.1
XM_006724269.3 Intron XP_006724332.1
XM_006724270.3 Intron XP_006724333.1
XM_011530239.2 Intron XP_011528541.1
XM_011530240.2 Intron XP_011528542.1
XM_011530241.2 Intron XP_011528543.1
XM_011530242.2 Intron XP_011528544.1
XM_011530243.2 Intron XP_011528545.1
XM_011530244.2 Intron XP_011528546.1
XM_011530245.2 Intron XP_011528547.1
XM_017028839.1 Intron XP_016884328.1
XM_017028840.1 Intron XP_016884329.1

View Full Product Details