Product Details

SNP ID
rs2241980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:208142897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATCTCTGTCGTAGATCTTCATT[C/G]TGTAAGCGCCAGAGTGCTGGAGTGG
Phenotype
MIM: 123670 MIM: 123680
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRYGB PubMed Links

Gene Details

Gene
CRYGB
Gene Name
crystallin gamma B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005210.3 302 Missense Mutation ACA,AGA T90R NP_005201.2
XM_017003402.1 302 Missense Mutation ACA,AGA T92R XP_016858891.1
Gene
CRYGC
Gene Name
crystallin gamma C
There are no transcripts associated with this gene.

Gene
LOC100507443
Gene Name
uncharacterized LOC100507443
There are no transcripts associated with this gene.

View Full Product Details