Product Details

SNP ID
rs2239006
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:119872086 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAGGGTGGTGGGCAGGGAGACCGT[T/C]AGCCTGCGAACCCTCTCTCCGAGGT
Phenotype
MIM: 300078 MIM: 300951
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
NDUFA1 PubMed Links

Gene Details

Gene
NDUFA1
Gene Name
NADH:ubiquinone oxidoreductase subunit A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004541.3 Intron NP_004532.1
Gene
RNF113A
Gene Name
ring finger protein 113A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006978.2 Intron NP_008909.1

View Full Product Details