Product Details

SNP ID
rs2242241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:21909370 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTCAGTTCCTGGCTGCCAGCCAG[C/A]AGCAGAGAAATCCTGCCCGGCTGGC
Phenotype
MIM: 604997
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
DOK2 PubMed Links

Gene Details

Gene
DOK2
Gene Name
docking protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317800.1 1199 Missense Mutation GCT,TCT A240S NP_001304729.1
NM_003974.3 1199 Missense Mutation GCT,TCT A394S NP_003965.2
NM_201349.2 1199 Missense Mutation GCT,TCT A240S NP_958728.1
XM_005273680.4 1199 Missense Mutation GCT,TCT A300S XP_005273737.1

View Full Product Details