Product Details

SNP ID
rs2239377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45508498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCAGAGGGCGGCTCCCCGCCTC[C/G]GGCCATAAGGGCGGCCCGGCTCCCC
Phenotype
MIM: 601703
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PPM1N PubMed Links

Gene Details

Gene
PPM1N
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1N (putative)
There are no transcripts associated with this gene.

Gene
VASP
Gene Name
vasodilator-stimulated phosphoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003370.3 Intron NP_003361.1
XM_005259199.1 Intron XP_005259256.1
XM_005259200.1 Intron XP_005259257.1
XM_017027200.1 Intron XP_016882689.1

View Full Product Details