Product Details

SNP ID
rs2240042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27202998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATGTCTGCATGTCCTGTCAGTA[A/T]TGAGTTGTAGCCACCAATCAGTGGA
Phenotype
MIM: 614060 MIM: 142959
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HOTTIP PubMed Links

Gene Details

Gene
HOTTIP
Gene Name
HOXA distal transcript antisense RNA
There are no transcripts associated with this gene.

Gene
HOXA13
Gene Name
homeobox A13
There are no transcripts associated with this gene.

View Full Product Details