Product Details

SNP ID
rs2266863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153503202 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACTGGGGAAGCATCGGGCAGGGC[A/G]GGGCTGCTCATAGGGTGCTGGCCTG
Phenotype
MIM: 301870 MIM: 300540
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BGN PubMed Links

Gene Details

Gene
BGN
Gene Name
biglycan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001711.5 Intron NP_001702.1
XM_017029724.1 Intron XP_016885213.1
Gene
HAUS7
Gene Name
HAUS augmin like complex subunit 7
There are no transcripts associated with this gene.

View Full Product Details