Product Details

SNP ID
rs2266928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:530124 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCTCAGCTGTGTGGAGCCACTGG[A/C]CTCTCATGCCCCACGCTGGAGAGAA
Phenotype
MIM: 603267 MIM: 608738
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAPN15 PubMed Links

Gene Details

Gene
CAPN15
Gene Name
calpain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005632.2 Intron NP_005623.1
XM_011522620.2 Intron XP_011520922.1
XM_011522621.2 Intron XP_011520923.1
XM_011522622.1 Intron XP_011520924.1
XM_011522623.2 Intron XP_011520925.1
XM_011522624.2 Intron XP_011520926.1
XM_011522625.2 Intron XP_011520927.1
XM_011522626.2 Intron XP_011520928.1
XM_011522627.2 Intron XP_011520929.1
XM_011522628.2 Intron XP_011520930.1
XM_011522629.2 Intron XP_011520931.1
XM_011522630.2 Intron XP_011520932.1
XM_011522631.2 Intron XP_011520933.1
XM_011522632.2 Intron XP_011520934.1
XM_017023596.1 Intron XP_016879085.1
Gene
LINC00235
Gene Name
long intergenic non-protein coding RNA 235
There are no transcripts associated with this gene.

Gene
MIR5587
Gene Name
microRNA 5587
There are no transcripts associated with this gene.

Gene
RAB11FIP3
Gene Name
RAB11 family interacting protein 3
There are no transcripts associated with this gene.

View Full Product Details