Product Details

SNP ID
rs2957859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61483598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCATATTTGAAGATGATAAGAGC[G/T]GGTGGAGTGCGGGGTGGGGCGGGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R32 PubMed Links

Gene Details

Gene
PPP1R32
Gene Name
protein phosphatase 1 regulatory subunit 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170753.1 Intron NP_001164224.1
NM_145017.2 Intron NP_659454.2

View Full Product Details