Product Details

SNP ID
rs2285634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13719600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAATCCACTACAGGTGAACTATGT[C/T]TTTATGTGCTTAAGCAACAGCAATT
Phenotype
MIM: 300284 MIM: 300202
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB9A PubMed Links

Gene Details

Gene
RAB9A
Gene Name
RAB9A, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TRAPPC2
Gene Name
trafficking protein particle complex 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011658.3 Intron NP_001011658.1
NM_001128835.2 Intron NP_001122307.2
NM_014563.5 Intron NP_055378.1
XM_011545565.1 Intron XP_011543867.1
XM_011545566.2 Intron XP_011543868.1

View Full Product Details