Product Details

SNP ID
rs2273058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:20052675 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGCATCCCCAGGTCAGCCTCCGGA[A/G]CTGGGATGACCAGGCGAGGACGAGT
Phenotype
MIM: 610952
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP61 PubMed Links

Gene Details

Gene
CFAP61
Gene Name
cilia and flagella associated protein 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167816.1 183 Intron NP_001161288.1
NM_015585.3 183 Intron NP_056400.3
XM_005260688.3 183 Intron XP_005260745.1
XM_005260689.2 183 Intron XP_005260746.1
XM_005260690.3 183 Intron XP_005260747.1
XM_005260691.3 183 Intron XP_005260748.1
XM_005260692.3 183 Intron XP_005260749.1
XM_005260693.3 183 Intron XP_005260750.1
XM_005260695.4 183 Intron XP_005260752.1
XM_005260696.4 183 Intron XP_005260753.1
XM_005260697.1 183 Intron XP_005260754.1
XM_011529211.1 183 Intron XP_011527513.1
XM_011529212.2 183 Intron XP_011527514.1
XM_011529213.1 183 Intron XP_011527515.1
XM_017027786.1 183 Intron XP_016883275.1
XM_017027787.1 183 Intron XP_016883276.1
XM_017027788.1 183 Intron XP_016883277.1
XM_017027789.1 183 Intron XP_016883278.1
XM_017027790.1 183 Intron XP_016883279.1
XM_017027791.1 183 Intron XP_016883280.1
XM_017027792.1 183 Intron XP_016883281.1
XM_017027793.1 183 Intron XP_016883282.1
XM_017027794.1 183 Intron XP_016883283.1
XM_017027795.1 183 Intron XP_016883284.1
Gene
CRNKL1
Gene Name
crooked neck pre-mRNA splicing factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278625.1 183 Missense Mutation CTC,TTC L39F NP_001265554.1
NM_001278626.1 183 UTR 5 NP_001265555.1
NM_001278627.1 183 Intron NP_001265556.1
NM_001278628.1 183 Intron NP_001265557.1
NM_016652.5 183 Missense Mutation CTC,TTC L51F NP_057736.4

View Full Product Details