Product Details

SNP ID
rs2285759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:109959410 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCTGGACTCAAACTTGCCAATCC[G/T]AGTGAAGGTTGTGGTTACATCCATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF3IP1 PubMed Links

Gene Details

Gene
EIF3IP1
Gene Name
eukaryotic translation initiation factor 3 subunit I pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details