Product Details

SNP ID
rs2273263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31619016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATCAGTGTCATCAGTGTCCTCA[C/G]AAGACACTAGCAGAGTCCAGGGTGA
Phenotype
MIM: 612770 MIM: 612765
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR7109 PubMed Links

Gene Details

Gene
MIR7109
Gene Name
microRNA 7109
There are no transcripts associated with this gene.

Gene
PISD
Gene Name
phosphatidylserine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001326411.1 2063 UTR 3 NP_001313340.1
NM_001326412.1 2063 UTR 3 NP_001313341.1
NM_001326413.1 2063 UTR 3 NP_001313342.1
NM_001326414.1 2063 UTR 3 NP_001313343.1
NM_001326415.1 2063 UTR 3 NP_001313344.1
NM_001326416.1 2063 UTR 3 NP_001313345.1
NM_001326417.1 2063 UTR 3 NP_001313346.1
NM_001326418.1 2063 UTR 3 NP_001313347.1
NM_001326419.1 2063 UTR 3 NP_001313348.1
NM_001326420.1 2063 UTR 3 NP_001313349.1
NM_001326421.1 2063 UTR 3 NP_001313350.1
NM_014338.3 2063 UTR 3 NP_055153.1
NM_178022.1 2063 UTR 3 NP_821141.1
Gene
SFI1
Gene Name
SFI1 centrin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007467.2 2063 Intron NP_001007468.1
NM_001258325.1 2063 Intron NP_001245254.1
NM_001258326.1 2063 Intron NP_001245255.1
NM_001258327.1 2063 Intron NP_001245256.1
NM_014775.3 2063 Intron NP_055590.2
XM_005261868.2 2063 Intron XP_005261925.1
XM_006724390.2 2063 Intron XP_006724453.1
XM_011530570.2 2063 Intron XP_011528872.1
XM_011530571.2 2063 Intron XP_011528873.1
XM_011530574.2 2063 Intron XP_011528876.1
XM_011530575.2 2063 Intron XP_011528877.1
XM_011530576.2 2063 Intron XP_011528878.1
XM_011530577.2 2063 Intron XP_011528879.1
XM_011530579.2 2063 Intron XP_011528881.1
XM_011530580.2 2063 Intron XP_011528882.1
XM_011530581.2 2063 Intron XP_011528883.1
XM_011530582.2 2063 Intron XP_011528884.1
XM_011530583.2 2063 Intron XP_011528885.1
XM_011530587.2 2063 Intron XP_011528889.1
XM_017029139.1 2063 Intron XP_016884628.1
XM_017029140.1 2063 Intron XP_016884629.1
XM_017029141.1 2063 Intron XP_016884630.1
XM_017029142.1 2063 Intron XP_016884631.1
XM_017029143.1 2063 Intron XP_016884632.1
XM_017029144.1 2063 Intron XP_016884633.1
XM_017029145.1 2063 Intron XP_016884634.1
XM_017029146.1 2063 Intron XP_016884635.1
XM_017029147.1 2063 Intron XP_016884636.1
XM_017029148.1 2063 Intron XP_016884637.1
XM_017029149.1 2063 Intron XP_016884638.1
XM_017029150.1 2063 Intron XP_016884639.1
XM_017029151.1 2063 Intron XP_016884640.1
XM_017029152.1 2063 Intron XP_016884641.1
XM_017029153.1 2063 Intron XP_016884642.1
XM_017029154.1 2063 Intron XP_016884643.1
XM_017029155.1 2063 Intron XP_016884644.1
XM_017029156.1 2063 Intron XP_016884645.1
XM_017029157.1 2063 Intron XP_016884646.1
XM_017029158.1 2063 Intron XP_016884647.1
XM_017029159.1 2063 Intron XP_016884648.1
XM_017029160.1 2063 Intron XP_016884649.1
XM_017029161.1 2063 Intron XP_016884650.1
XM_017029162.1 2063 Intron XP_016884651.1

View Full Product Details