Product Details

SNP ID
rs2273262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31618998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCAAAGGTCTGTATCATTAATCA[A/G]TGTCATCAGTGTCCTCAGAAGACAC
Phenotype
MIM: 612770 MIM: 612765
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR7109 PubMed Links

Gene Details

Gene
MIR7109
Gene Name
microRNA 7109
There are no transcripts associated with this gene.

Gene
PISD
Gene Name
phosphatidylserine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001326411.1 2081 UTR 3 NP_001313340.1
NM_001326412.1 2081 UTR 3 NP_001313341.1
NM_001326413.1 2081 UTR 3 NP_001313342.1
NM_001326414.1 2081 UTR 3 NP_001313343.1
NM_001326415.1 2081 UTR 3 NP_001313344.1
NM_001326416.1 2081 UTR 3 NP_001313345.1
NM_001326417.1 2081 UTR 3 NP_001313346.1
NM_001326418.1 2081 UTR 3 NP_001313347.1
NM_001326419.1 2081 UTR 3 NP_001313348.1
NM_001326420.1 2081 UTR 3 NP_001313349.1
NM_001326421.1 2081 UTR 3 NP_001313350.1
NM_014338.3 2081 UTR 3 NP_055153.1
NM_178022.1 2081 UTR 3 NP_821141.1
Gene
SFI1
Gene Name
SFI1 centrin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007467.2 2081 Intron NP_001007468.1
NM_001258325.1 2081 Intron NP_001245254.1
NM_001258326.1 2081 Intron NP_001245255.1
NM_001258327.1 2081 Intron NP_001245256.1
NM_014775.3 2081 Intron NP_055590.2
XM_005261868.2 2081 Intron XP_005261925.1
XM_006724390.2 2081 Intron XP_006724453.1
XM_011530570.2 2081 Intron XP_011528872.1
XM_011530571.2 2081 Intron XP_011528873.1
XM_011530574.2 2081 Intron XP_011528876.1
XM_011530575.2 2081 Intron XP_011528877.1
XM_011530576.2 2081 Intron XP_011528878.1
XM_011530577.2 2081 Intron XP_011528879.1
XM_011530579.2 2081 Intron XP_011528881.1
XM_011530580.2 2081 Intron XP_011528882.1
XM_011530581.2 2081 Intron XP_011528883.1
XM_011530582.2 2081 Intron XP_011528884.1
XM_011530583.2 2081 Intron XP_011528885.1
XM_011530587.2 2081 Intron XP_011528889.1
XM_017029139.1 2081 Intron XP_016884628.1
XM_017029140.1 2081 Intron XP_016884629.1
XM_017029141.1 2081 Intron XP_016884630.1
XM_017029142.1 2081 Intron XP_016884631.1
XM_017029143.1 2081 Intron XP_016884632.1
XM_017029144.1 2081 Intron XP_016884633.1
XM_017029145.1 2081 Intron XP_016884634.1
XM_017029146.1 2081 Intron XP_016884635.1
XM_017029147.1 2081 Intron XP_016884636.1
XM_017029148.1 2081 Intron XP_016884637.1
XM_017029149.1 2081 Intron XP_016884638.1
XM_017029150.1 2081 Intron XP_016884639.1
XM_017029151.1 2081 Intron XP_016884640.1
XM_017029152.1 2081 Intron XP_016884641.1
XM_017029153.1 2081 Intron XP_016884642.1
XM_017029154.1 2081 Intron XP_016884643.1
XM_017029155.1 2081 Intron XP_016884644.1
XM_017029156.1 2081 Intron XP_016884645.1
XM_017029157.1 2081 Intron XP_016884646.1
XM_017029158.1 2081 Intron XP_016884647.1
XM_017029159.1 2081 Intron XP_016884648.1
XM_017029160.1 2081 Intron XP_016884649.1
XM_017029161.1 2081 Intron XP_016884650.1
XM_017029162.1 2081 Intron XP_016884651.1

View Full Product Details