Product Details

SNP ID
rs2273341
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:11056424 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CCAGCTTGTGAATCTGGGCAGTCGC[C/T]TGGCTCCTGCCTACTGTCCTGAGCC
Phenotype
MIM: 605102 MIM: 182891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MASP2 PubMed Links

Gene Details

Gene
MASP2
Gene Name
mannan binding lectin serine peptidase 2
There are no transcripts associated with this gene.

Gene
SRM
Gene Name
spermidine synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003132.2 Intron NP_003123.2
XM_017002179.1 Intron XP_016857668.1

View Full Product Details