Product Details

SNP ID
rs2273853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39908657 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGGGCTCATGGTCCAGATACACC[A/G]TGCATGGATCATTGGGTACTACAGA
Phenotype
MIM: 606627 MIM: 603707
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAAM2 PubMed Links

Gene Details

Gene
DAAM2
Gene Name
dishevelled associated activator of morphogenesis 2
There are no transcripts associated with this gene.

Gene
MOCS1
Gene Name
molybdenum cofactor synthesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001075098.3 Intron NP_001068566.1
NM_005943.5 Intron NP_005934.2
XM_011514632.2 Intron XP_011512934.1
XM_011514633.2 Intron XP_011512935.1
XM_011514634.2 Intron XP_011512936.1
XM_017010888.1 Intron XP_016866377.1

View Full Product Details