Product Details

SNP ID
rs2286612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64226758 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCCCATGACCGTCGCCCACTGCCA[G/T]GTGGGGACGAGGCCATCACTGCCAT
Phenotype
MIM: 604189 MIM: 607901 MIM: 610470 MIM: 601398
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
There are no transcripts associated with this gene.

Gene
FERMT3
Gene Name
fermitin family member 3
There are no transcripts associated with this gene.

Gene
NUDT22
Gene Name
nudix hydrolase 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128612.2 164 Missense Mutation GGT,TGT G36C NP_001122084.1
NM_001128613.2 164 Missense Mutation GGT,TGT G36C NP_001122085.1
NM_001271831.1 164 Missense Mutation GGT,TGT G36C NP_001258760.1
NM_032344.3 164 Missense Mutation GGT,TGT G36C NP_115720.1
Gene
TRPT1
Gene Name
tRNA phosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033678.3 164 Intron NP_001028850.2
NM_001160389.1 164 Intron NP_001153861.1
NM_001160390.1 164 Intron NP_001153862.1
NM_001160392.1 164 Intron NP_001153864.1
NM_001160393.1 164 Intron NP_001153865.1
NM_031472.3 164 Intron NP_113660.1
XM_005274345.4 164 Intron XP_005274402.1
XM_005274346.4 164 Intron XP_005274403.1
XM_005274347.4 164 Intron XP_005274404.1
XM_006718706.3 164 Intron XP_006718769.1
XM_006718707.3 164 Intron XP_006718770.1
XM_017018400.1 164 Intron XP_016873889.1
XM_017018401.1 164 Intron XP_016873890.1
Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details