Product Details

SNP ID
rs2273937
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:227732802 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTAAGACAGAGGCACTGAGAAGCTG[C/T]GCGGTGACCAAGGGAGTCTGAGCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JMJD4 PubMed Links

Gene Details

Gene
JMJD4
Gene Name
jumonji domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161465.1 652 Intron NP_001154937.1
NM_023007.2 652 Intron NP_075383.2
XM_011544262.2 652 Intron XP_011542564.1
Gene
SNAP47
Gene Name
synaptosome associated protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323930.1 652 Intron NP_001310859.1
NM_001323931.1 652 Intron NP_001310860.1
NM_001323932.1 652 Intron NP_001310861.1
NM_001323933.1 652 Intron NP_001310862.1
NM_001323934.1 652 Intron NP_001310863.1
NM_001323935.1 652 Intron NP_001310864.1
NM_053052.3 652 Intron NP_444280.2
XM_006711734.3 652 Intron XP_006711797.1
XM_017000231.1 652 UTR 5 XP_016855720.1
XM_017000232.1 652 UTR 5 XP_016855721.1

View Full Product Details