Product Details

SNP ID
rs2274187
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:63323280 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CCCATCACGGACAGCCTCAGCCGCC[A/C]CACCAGCAGCCCCTGACATTGCCCA
Phenotype
MIM: 611539
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FOXD3 PubMed Links

Gene Details

Gene
FOXD3
Gene Name
forkhead box D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012183.2 222 Silent Mutation CCA,CCC P74P NP_036315.1
Gene
FOXD3-AS1
Gene Name
FOXD3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
LINC00466
Gene Name
long intergenic non-protein coding RNA 466
There are no transcripts associated with this gene.

Gene
MIR6068
Gene Name
microRNA 6068
There are no transcripts associated with this gene.

View Full Product Details